Learn More
Medchemexpress LLC TCO-PNB ester | 1438415-89-4 | 98.5% | C15H17NO5 | 100 MG
SDP
Catalog No. 5000353400
Encompass_Preferred
TCO-PNB ester is an amine-reactive building block used for the modification of amine-containing molecules. It contains a TCO group that can undergo an inverse electron demand Diels-Alder reaction (iEDDA) with molecules containing a Tetrazine group.
  • Amine-reactive building block
  • Used for modification of amine-containing molecules
  • Contains a TCO group
  • Undergoes inverse electron demand Diels-Alder reaction (iEDDA)
  • Reacts with molecules containing a Tetrazine group
Catalog No. 50-003-53400 Supplier Medchemexpress LLC Supplier No. HY141175100MG
TCO-PNB ester is an amine-reactive building block used for the modification of amine-containing molecules. It contains a TCO group that can undergo an inverse electron demand Diels-Alder reaction (iEDDA) with molecules containing a Tetrazine group.
  • Amine-reactive building block
  • Used for modification of amine-containing molecules
  • Contains a TCO group
  • Undergoes inverse electron demand Diels-Alder reaction (iEDDA)
  • Reacts with molecules containing a Tetrazine group