Learn More
Medchemexpress LLC TCO-PNB ester | 1438415-89-4 | 98.5% | C15H17NO5 | 25 MG
SDP
Catalog No. 5000352493
Encompass_Preferred
TCO-PNB ester is an amine-reactive building block used for the modification of amine-containing molecules. It contains a TCO group that can undergo an inverse electron demand Diels-Alder reaction (iEDDA) with molecules containing a Tetrazine group.
  • Amine-reactive building block for modification of amine-containing molecules.
  • The TCO group allows for iEDDA reaction with Tetrazine-containing molecules.
Catalog No. 50-003-52493 Supplier Medchemexpress LLC Supplier No. 25MGHY141175
TCO-PNB ester is an amine-reactive building block used for the modification of amine-containing molecules. It contains a TCO group that can undergo an inverse electron demand Diels-Alder reaction (iEDDA) with molecules containing a Tetrazine group.
  • Amine-reactive building block for modification of amine-containing molecules.
  • The TCO group allows for iEDDA reaction with Tetrazine-containing molecules.