Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tco-Pnb Ester

Catalog No. 90092750MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-092-750MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-092-750MG Supplier Sigma Aldrich Supplier No. 90092750MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

TCO-PNB ester (trans-Cyclooctene-p-nitrobenzyl ester) has a p-nitrobenzyl (PNB) ester group that is amine reactive and a trans-cyclooctene (TCO) group that is reactive with tetrazines. TCO-PNB ester is supplied as a single axial (major) diastereomer. This specially formulated crystalline material provides easy handling and extended shelf life.

Specifications

CAS 1339202-33-3
Molecular Formula C7H13BrO4
Linear Formula C7H13BrO4
MDL Number MFCD28122969
Quantity 50 mg
Molecular Weight (g/mol) 241.08 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.