Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC TCO-SS-amine | >=95.0% | 304.47 | C13H24N2O2S2 | 5 MG
SDP

Catalog No. 5000203725
Encompass_Preferred

TCO-SS-amine is a cleavable antibody-drug conjugate (ADC) linker containing a trans-cyclooctene (TCO) group that enables inverse electron demand Diels-Alder (iEDDA) click reactions with tetrazine-functionalized partners. It is used as a disulfide-cleavable linker in ADC synthesis and as a bioorthogonal click chemistry reagent.

  • Cleavable disulfide linker for ADC synthesis.
  • Contains a TCO group for iEDDA click chemistry with tetrazines.
  • Molecular formula: C13H24N2O2S2.
  • Molecular weight: 304.47 g/mol.
  • Purity: ≥95.0%.
  • Storage: -20°C, protect from light; in solvent: -80°C up to 6 months.
  • Available in small research pack sizes such as 5 mg.

Catalog No. 50-002-03725 Supplier Medchemexpress LLC Supplier No. HY1360395MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TCO-SS-amine is a cleavable antibody-drug conjugate (ADC) linker containing a trans-cyclooctene (TCO) group that enables inverse electron demand Diels-Alder (iEDDA) click reactions with tetrazine-functionalized partners. It is used as a disulfide-cleavable linker in ADC synthesis and as a bioorthogonal click chemistry reagent.

  • Cleavable disulfide linker for ADC synthesis.
  • Contains a TCO group for iEDDA click chemistry with tetrazines.
  • Molecular formula: C13H24N2O2S2.
  • Molecular weight: 304.47 g/mol.
  • Purity: ≥95.0%.
  • Storage: -20°C, protect from light; in solvent: -80°C up to 6 months.
  • Available in small research pack sizes such as 5 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.