Learn More
Enzo Life Sciences TCP1 alpha monoclonal antibody (23c) (200µg)
SDP
Supplier:  Enzo Life Sciences ADICTA123F
Encompass_Preferred
Clone: 23c. Host: Rat. Isotype: IgG2c. Immunogen: Recombinant mouse TCP-1α (C-terminal fragment). UniProt ID: P11983. Source: Purified from mouse ascites. Species reactivity: Human, Mouse, Rat, Sheep, Rabbit, Plant, Hamster, Dog, Chicken. Applications: ICC, IHC, IP, WB. Recommended Dilutions: Western Blot (1:1,000, colorimetric) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7206