Learn More
Enzo Life Sciences TCP1 alpha monoclonal antibody (91a) (50µg)
SDP
Supplier:  Enzo Life Sciences ADICTA191D
Encompass_Preferred
Clone: 91a. Host: Rat. Isotype: IgG2a. Immunogen: Recombinant mouse TCP-1α (C-terminal fragment). Source: Purified from mouse ascites. Species reactivity: Human, Mouse, Rat, Yeast, Rabbit, Plant, Porcine, Monkey, Hamster, Guinea pig, Drosophila, Dog, C. elegans, Bovine. Applications: Flow Cytometry, ICC, IHC, IP, WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7207