Learn More
Aviva Systems Biology Corp TCP1 Antibody - C-terminal region (ARP63619_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63619_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TCP1. It was validated on Western Blot. . Alternate names: CCT-alpha, CCT1, CCTa, D6S230E, TCP-1-alphaApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-288-035