Learn More
Enzo Life Sciences TCPOBOP (25mg). CAS: 76150-91-9
SDP
Supplier:  Enzo Life Sciences BMLGR2450025
Encompass_Preferred
TCPOBOP is an activator of the mouse constitutive androstane receptor (CAR) and a potent inducer of cytochrome P450 monoxygenase activity. Has been shown to have a potent tumor-promoting activity and hepatocarcinogenic effects in mice but not in rats. Alternative name: 1,4-Bis-[2-(3,5-dichloropyridyloxy)]benzene. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (5mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-0949