Learn More
Abcam TCPOBOP, Androstane receptor (mCAR) agonist, 50MG
Catalog No. 502706709
Encompass_Preferred
MW 402.1 Da, Purity >99%. Potent murine constitutive androstane receptor (mCAR) agonist. Pharmacologically active in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6709 Supplier Abcam Supplier No. AB14129450MG
Add to Cart
Add to Cart
MW 402.1 Da, Purity >99%. Potent murine constitutive androstane receptor (mCAR) agonist. Pharmacologically active in vivo.
The product is subject to the following: Abcam Restricted Use Statement