Learn More
Eurofins Discoverx TCPTP, 10 µg. (14-646)
Supplier:  Eurofins Discoverx 14646
Encompass
The TCPTP product is purified is untagged recombinant TCPTP enzyme. This enzyme contains amino acids 1-341. It retains its binding and enzymatic activity and is intended for use in phosphatase assays. This enzyme is provided as 10 μg of 0.187 mg/mL frozen solution. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9113