Learn More
Enzo Life Sciences T cell protein tyrosine phosphatase (human), (recombinant) (200 U)
SDP
Supplier:  Enzo Life Sciences BMLSE1140200
Encompass_Preferred
Alternative name: TCPTP, T cell PTP. Purity: ≥95% (SDS-PAGE). Formulation: Liquid. In 50mM Tris-HCl, pH 7.0 containing 100 mM NaCl, 2 mM EDTA, 5 mM DTT, 0.01% Brij35 and 30% glycerol. Source: Produced in E. coli. T-cell phosphatase with a C-terminal 11 kDa deletion (TCΔC11) (aa 1-317). Long Term Storage: -80°C.
Catalog No. 50-201-1934