Learn More
CAPRICO BIOTECHNOLOGIES INC TCR A/B FITC, 4220015
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4220015
Encompass_Preferred
The TCR A/B FITC Antibody is a monoclonal antibody that recognizes the T cell receptor alpha/beta heterodimer. It is conjugated to FITC and is suitable for flow cytometry, immunofluorescence microscopy, and immunohistochemistry. This antibody is produced in mice and recognizes human, mouse, and rat TCR A/B. It can be used to identify TCR A/B expressing cells in various research applications. The antibody is supplied as 100 tests in PBS with 0.1% sodium azide and 0.2% BSA.
Catalog No. 50-227-9322