Learn More
Beckman Coulter Beckman Coulter TCR Vβ11-APC Antibody, Clone C21, 0.5 mL, ASR (A66905)
Supplier: Beckman Coulter A66905
Beckman Coulter TCR Vβ11-APC antibody (clone C21) is an APC-conjugated monoclonal antibody for detection of the human T cell receptor Vβ11 region by flow cytometry. Supplied as a 0.5 mL ASR reagent; analytical and performance characteristics are not established.
- Size: 0.5 mL.
- Conjugate: APC.
- Clone: C21.
- Target: human TCR Vβ11 (TRBV25-1).
- Host species: mouse.
- Isotype: IgG2a.
- Regulatory status: ASR; analytical and performance characteristics not established.
- Intended use: detection of TCR Vβ11 by flow cytometry.
Description
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.