Learn More
Beckman Coulter TCR Vβ11-PE, C21, 1 mL, ASR
Supplier:  Beckman Coulter IM2290
Encompass
Also called TCRBV11S1 Vβ11 is the human variable β11 chain of the T cell receptor. Two Vβ11 sequences are described, PL3.12 and PH15. These sequences differ only in their leader sequence and therefore lead to the same mature protein.
Catalog No. NC1779574