Learn More
American Research Products Inc TCRcure A06 Biosimilar
Catalog No. 5000306937
Encompass
TCRcure A06 Biosimilar (Anti-ALPP) Antibody is a biosimilar antibody with human reactivity produced in humanized Available in 100 ug size with 7 - 14 business days lead time Ships via gel pack Store at -20 C to -80 C for 12 months in lyophilized form After reconstitution if not intended for use within a month aliquot and store at -80 C (Avoid repeated freezing and thawing) Lyophilized antibodies are shipped at ambient temperature
Catalog No. 50-003-06937 Supplier American Research Products Inc Supplier No. HDBS0149100UG
TCRcure A06 Biosimilar (Anti-ALPP) Antibody is a biosimilar antibody with human reactivity produced in humanized Available in 100 ug size with 7 - 14 business days lead time Ships via gel pack Store at -20 C to -80 C for 12 months in lyophilized form After reconstitution if not intended for use within a month aliquot and store at -80 C (Avoid repeated freezing and thawing) Lyophilized antibodies are shipped at ambient temperature