Learn More
INVIVOSCRIBE TECHNOLOGIES IdentiClone T-cell Receptor Gamma Gene Rearrangement Assay 2.0, PCR-based, T-cell Receptor Gamma Gene Rearrangements, Clonality detection in lymphoproliferations, ABI Fluorescence, 33 rxns, Research Use Only

Supplier: INVIVOSCRIBE TECHNOLOGIES 12070101
This IdentiClone T-cell Receptor Gamma Gene Rearrangement Assay 2.0 from Invivoscribe Technologies is a PCR-based assay designed for the precise detection of clonal T-cell receptor gamma gene rearrangements. Utilizing ABI Fluorescence for detection, this assay provides a robust method for identifying clonality in patients with suspected lymphoproliferations, aiding in the investigation of lymphoid malignancies.
- Assay type: PCR-based
- Target: T-cell receptor gamma gene rearrangements
- Application: Clonality detection in lymphoproliferations
- Detection method: ABI Fluorescence
- Number of reactions: 33 rxns
- Usage: Research use only
- Supplier: Invivoscribe Technologies
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.