Learn More
TARGETMOL CHEMICALS INC TCS 2210 5MG
SDP
Catalog No. 502580869
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. TCS 2210 is an inducer of neuronal differentiation in mesenchymal stem cells and increases the neuronal markers (beta)-III tubulin expression and neuron-specific enolase expression. purity: 97%
Catalog No. 50-258-0869 Supplier TARGETMOL CHEMICALS INC Supplier No. T234465MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. TCS 2210 is an inducer of neuronal differentiation in mesenchymal stem cells and increases the neuronal markers (beta)-III tubulin expression and neuron-specific enolase expression. purity: 97%