Learn More
Medchemexpress LLC HY-12308 5mg Medchemexpress, TCS 2314 CAS:317353-73-4 Purity:>98%
SDP
Catalog No. 501969644
Encompass_Preferred
Medchemexpress, HY-12308 5mg TCS 2314 CAS:317353-73-4 TCS 2314 (compound 3) is orally active and selective very late antigen-4 (VLA-4, α4β1, CD49d/CD29) antagonist with an IC50 of 4.4 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9644 Supplier Medchemexpress LLC Supplier No. HY123085MG
Medchemexpress, HY-12308 5mg TCS 2314 CAS:317353-73-4 TCS 2314 (compound 3) is orally active and selective very late antigen-4 (VLA-4, α4β1, CD49d/CD29) antagonist with an IC50 of 4.4 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.