Learn More
Medchemexpress LLC TCS 2510 | 346673-06-1 | 98.3% | 383.49 | 1 ML
SDP
Catalog No. 5000346970
Encompass_Preferred
TCS 2510 is a selective EP4 agonist. It can be used for the research of metabolic diseases. This product is for research use only and not sold to patients.
  • Stimulates glucagon like peptide 1 secretion in GLUTag cells.
  • Activates the Gas signaling pathway.
  • Increases Gcg mRNA expression levels in GLUTag cells.
  • Produces a concentration-dependent increase in cAMP levels.
  • Elevates plasma glucagon like peptide 1 levels in fasted mice.
Catalog No. 50-003-46970 Supplier Medchemexpress LLC Supplier No. HY1085571MG
TCS 2510 is a selective EP4 agonist. It can be used for the research of metabolic diseases. This product is for research use only and not sold to patients.
  • Stimulates glucagon like peptide 1 secretion in GLUTag cells.
  • Activates the Gas signaling pathway.
  • Increases Gcg mRNA expression levels in GLUTag cells.
  • Produces a concentration-dependent increase in cAMP levels.
  • Elevates plasma glucagon like peptide 1 levels in fasted mice.