Learn More
Medchemexpress LLC TCS 2510 | 346673-06-1 | 98.3% | 383.49 | 10 MG
SDP
Catalog No. 5000346535
Encompass_Preferred
TCS 2510 is a selective EP4 agonist. It is used for the research of metabolic diseases.
  • Stimulates glucagon like peptide 1 secretion.
  • Activates the Gas signaling pathway.
  • Increases Gcg mRNA expression levels in GLUTag cells.
  • Produces a concentration dependent increase in cAMP levels in GLUTag cells.
  • Decreases cell impedance in GLUTag cells in a concentration dependent manner.
  • Produces a concentration-dependent increase in glucagon like peptide 2 secretion in GLUTag cells.
  • Elevates plasma glucagon like peptide 1 levels in fasted mice.
  • Increases plasma glucagon like peptide 2 levels.
Catalog No. 50-003-46535 Supplier Medchemexpress LLC Supplier No. HY10855710MG
TCS 2510 is a selective EP4 agonist. It is used for the research of metabolic diseases.
  • Stimulates glucagon like peptide 1 secretion.
  • Activates the Gas signaling pathway.
  • Increases Gcg mRNA expression levels in GLUTag cells.
  • Produces a concentration dependent increase in cAMP levels in GLUTag cells.
  • Decreases cell impedance in GLUTag cells in a concentration dependent manner.
  • Produces a concentration-dependent increase in glucagon like peptide 2 secretion in GLUTag cells.
  • Elevates plasma glucagon like peptide 1 levels in fasted mice.
  • Increases plasma glucagon like peptide 2 levels.