Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC TCS 359 | 301305-73-7 | 99.8% | 360.43 | 1 ML
SDP

Catalog No. 5000348360
Encompass_Preferred

TCS 359 is a 2-acylaminothiophene-3-carboxamide, functioning as a potent and selective FLT3 inhibitor with an IC50 of 42 nM. It demonstrates inhibition of MV4-11 cell proliferation with an IC50 of 340 nM. This compound is suitable for laboratory research and substance manufacturing.

  • Potent and selective FLT3 inhibitor (IC50 of 42 nM)
  • Inhibits MV4-11 cell proliferation (IC50 of 340 nM)
  • High purity of 99.76%
  • Solid appearance, white to yellow in color
  • Recommended storage at 4°C, protected from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light
  • Causes skin irritation
  • Causes serious eye irritation
  • May cause respiratory irritation

Catalog No. 50-003-48360 Supplier Medchemexpress LLC Supplier No. HY1390710MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TCS 359 is a 2-acylaminothiophene-3-carboxamide, functioning as a potent and selective FLT3 inhibitor with an IC50 of 42 nM. It demonstrates inhibition of MV4-11 cell proliferation with an IC50 of 340 nM. This compound is suitable for laboratory research and substance manufacturing.

  • Potent and selective FLT3 inhibitor (IC50 of 42 nM)
  • Inhibits MV4-11 cell proliferation (IC50 of 340 nM)
  • High purity of 99.76%
  • Solid appearance, white to yellow in color
  • Recommended storage at 4°C, protected from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light
  • Causes skin irritation
  • Causes serious eye irritation
  • May cause respiratory irritation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.