Learn More
Abcam TCS 359, FLT3 receptor tyrosine kinase inhibitor, 10MG
Catalog No. 502706738
Encompass_Preferred
MW 360.4 Da, Purity >99%. Potent FLT3 receptor tyrosine kinase inhibitor (IC₅₀ = 42 nM). Selective over 22 other kinases. Shows antiproliferative effects (IC₅₀ = 340 nM for Human acute myelocytic leukemia MV4-11 cells).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6738 Supplier Abcam Supplier No. AB14126810MG
MW 360.4 Da, Purity >99%. Potent FLT3 receptor tyrosine kinase inhibitor (IC₅₀ = 42 nM). Selective over 22 other kinases. Shows antiproliferative effects (IC₅₀ = 340 nM for Human acute myelocytic leukemia MV4-11 cells).
The product is subject to the following: Abcam Restricted Use Statement