Learn More
Cayman Chemical TCS 5861528, Purity: ≥98%, CAS: 332117-28-9, 50 mg
SDP
Catalog No. 502979531
Encompass_Preferred
A selective TRPA1 blocker that antagonizes TRPA1-mediated calcium influx induced by allyl isothiocyanate and 4-hydroxy nonenal (IC50s = 14.3 and 18.7 μM, respectively); 30 mg/kg twice daily in rats attenuates pain-related behavior in models of diabetic hypersensitivity and postoperative pain
Catalog No. 50-297-9531 Supplier Cayman Chemical Supplier No. 1838050 MG
A selective TRPA1 blocker that antagonizes TRPA1-mediated calcium influx induced by allyl isothiocyanate and 4-hydroxy nonenal (IC50s = 14.3 and 18.7 μM, respectively); 30 mg/kg twice daily in rats attenuates pain-related behavior in models of diabetic hypersensitivity and postoperative pain