Learn More
Cayman Chemical TCS ERK 11e, Purity: ≥98%, CAS: 896720-20-0, 10 mg
SDP
Catalog No. 502981503
Encompass_Preferred
A selective inhibitor of ERK2 (Ki 50 = 48 nM)
Catalog No. 50-298-1503 Supplier Cayman Chemical Supplier No. 1993210 MG
A selective inhibitor of ERK2 (Ki 50 = 48 nM)