Learn More
Cayman Chemical TCS ERK 11e, Purity: ≥98%, CAS: 896720-20-0, 25 mg
SDP
Catalog No. 502985541
Encompass_Preferred
A selective inhibitor of ERK2 (Ki 50 = 48 nM)
Catalog No. 50-298-5541 Supplier Cayman Chemical Supplier No. 1993225 MG
A selective inhibitor of ERK2 (Ki 50 = 48 nM)