Learn More
Medchemexpress LLC TCS-OX2-29 | 372523-75-6 | 99.6% | 397.51 g/mol | C23H31N3O3 | 25 MG
SDP
Catalog No. 5000214123
Encompass_Preferred
TCS-OX2-29 is a potent, selective orexin-2 receptor (OX2R) antagonist used in neuropharmacology research to probe orexin signaling and sleep-wake regulation. Reported activity includes an IC50 of 40 nM and a pKi of 7.5, with approximately 250-fold selectivity for OX2 over OX1. The compound is supplied as a high-purity solid and is available in DMSO solution formats for easy reconstitution.
  • Potent OX2R antagonist with IC50 ≈ 40 nM.
  • High selectivity for OX2 over OX1 (~250-fold).
  • High purity suitable for research applications (≈99.6%).
  • Available as a solid and as 10 mM DMSO solution for reconstitution.
  • Molecular weight 397.51 g/mol; molecular formula C23H31N3O3.
Catalog No. 50-002-14123 Supplier Medchemexpress LLC Supplier No. HY10045225MG
TCS-OX2-29 is a potent, selective orexin-2 receptor (OX2R) antagonist used in neuropharmacology research to probe orexin signaling and sleep-wake regulation. Reported activity includes an IC50 of 40 nM and a pKi of 7.5, with approximately 250-fold selectivity for OX2 over OX1. The compound is supplied as a high-purity solid and is available in DMSO solution formats for easy reconstitution.
  • Potent OX2R antagonist with IC50 ≈ 40 nM.
  • High selectivity for OX2 over OX1 (~250-fold).
  • High purity suitable for research applications (≈99.6%).
  • Available as a solid and as 10 mM DMSO solution for reconstitution.
  • Molecular weight 397.51 g/mol; molecular formula C23H31N3O3.