Learn More
Cayman Chemical TCS PIM-1 1, Purity: ≥98%, CAS: 491871-58-0, 5 mg
SDP
Catalog No. 502970396
Encompass_Preferred
An ATP-competitive Pim-1 kinase inhibitor (IC50 = 50 nM) that displays selectivity over the related kinases, Pim-2 and MEK1/2 (IC50s = >20 µM)
Catalog No. 50-297-0396 Supplier Cayman Chemical Supplier No. 162865 MG
An ATP-competitive Pim-1 kinase inhibitor (IC50 = 50 nM) that displays selectivity over the related kinases, Pim-2 and MEK1/2 (IC50s = >20 µM)