Learn More
Medchemexpress LLC TCS PrP Inhibitor 13 25mg | 34320-83-7 | 25MG
SDP
Catalog No. 5000241588
Encompass_Preferred
TCS PrP Inhibitor 13 an antiprion agent is a cellular prion protein (PrPC) inhibitor TCS PrP Inhibitor 13 as a protease-resistant form of prion protein (PrP-res) accumulation inhibitor shows an IC50 value of 3 nM in both ScN2a and F3 cell lines TCS PrP Inhibitor 13 induces Schwannoma cells apoptosis[1
Catalog No. 50-002-41588 Supplier Medchemexpress LLC Supplier No. HY10766225MG
TCS PrP Inhibitor 13 an antiprion agent is a cellular prion protein (PrPC) inhibitor TCS PrP Inhibitor 13 as a protease-resistant form of prion protein (PrP-res) accumulation inhibitor shows an IC50 value of 3 nM in both ScN2a and F3 cell lines TCS PrP Inhibitor 13 induces Schwannoma cells apoptosis[1