Learn More
Medchemexpress LLC TCS7010 | 1158838-45-9 | 99.8% | C31H31ClFN7O2 | 250 MG
SDP
Catalog No. 5000358717
Encompass_Preferred
TCS7010 is a potent and highly selective Aurora A inhibitor with an IC50 of 3.4 nM. It is considered a useful tool for investigating the cellular role of Aurora A kinases without the complication of also inhibiting Aurora B.
  • Potent and highly selective Aurora A inhibitor
  • IC50 of 3.4 nM for Aurora A
  • Useful tool for investigating cellular role of Aurora A kinases
  • Does not inhibit Aurora B at relevant concentrations
Catalog No. 50-003-58717 Supplier Medchemexpress LLC Supplier No. 250MGHY70061
TCS7010 is a potent and highly selective Aurora A inhibitor with an IC50 of 3.4 nM. It is considered a useful tool for investigating the cellular role of Aurora A kinases without the complication of also inhibiting Aurora B.
  • Potent and highly selective Aurora A inhibitor
  • IC50 of 3.4 nM for Aurora A
  • Useful tool for investigating cellular role of Aurora A kinases
  • Does not inhibit Aurora B at relevant concentrations