Learn More
Medchemexpress LLC HY-19121A 5mg Medchemexpress, TCV-309 (chloride) CAS:121494-09-5 Purity:>98%
SDP
Catalog No. 502082108
Encompass_Preferred
Medchemexpress, HY-19121A 5mg TCV-309 (chloride) CAS:121494-09-5 TCV-309 chloride is a potent and specific platelet activating factor (PAF) antagonist. TCV-309 chloride specifically inhibits PAF-induced aggregation of rabbit and human platelets, and [3H]PAF binding to rabbit platelet microsomes with IC50 values of 33 nM, 58 nM and 27 nM, respectively. TCV-309 chloride has beneficial effects in anaphylactic shock. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2108 Supplier Medchemexpress LLC Supplier No. HY19121A5MG
Medchemexpress, HY-19121A 5mg TCV-309 (chloride) CAS:121494-09-5 TCV-309 chloride is a potent and specific platelet activating factor (PAF) antagonist. TCV-309 chloride specifically inhibits PAF-induced aggregation of rabbit and human platelets, and [3H]PAF binding to rabbit platelet microsomes with IC50 values of 33 nM, 58 nM and 27 nM, respectively. TCV-309 chloride has beneficial effects in anaphylactic shock. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.