Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC TD-0212 TFA 10mg | 1073549-11-7 | 10 MG
SDP

Catalog No. 5000229064
Encompass_Preferred

TD-0212 TFA is a research-grade small molecule supplied as the trifluoroacetate (TFA) salt that acts as an orally active dual angiotensin II type 1 (AT1) receptor antagonist and neprilysin (NEP) inhibitor. It is provided as a white to off-white solid for laboratory research use only.

  • Dual AT1 antagonist and NEP inhibitor with reported pKi 8.9 and pIC50 9.2.
  • High purity (98.44%) suitable for biological research.
  • Molecular formula C30H35F4N3O6S; molecular weight 641.67.
  • Soluble in DMSO (125 mg/mL) and compatible with in vivo co-solvent preparation protocols.
  • Stable when stored sealed at -20°C; in solution store at -80°C for long term.

Catalog No. 50-002-29064 Supplier Medchemexpress LLC Supplier No. HY114412A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TD-0212 TFA is a research-grade small molecule supplied as the trifluoroacetate (TFA) salt that acts as an orally active dual angiotensin II type 1 (AT1) receptor antagonist and neprilysin (NEP) inhibitor. It is provided as a white to off-white solid for laboratory research use only.

  • Dual AT1 antagonist and NEP inhibitor with reported pKi 8.9 and pIC50 9.2.
  • High purity (98.44%) suitable for biological research.
  • Molecular formula C30H35F4N3O6S; molecular weight 641.67.
  • Soluble in DMSO (125 mg/mL) and compatible with in vivo co-solvent preparation protocols.
  • Stable when stored sealed at -20°C; in solution store at -80°C for long term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.