Learn More
Medchemexpress LLC TD-0212 TFA 10mM | 10MM 1ML
SDP
Catalog No. 5000230011
Encompass_Preferred
TD-0212 TFA is an orally active dual pharmacology angiotensin II type 1 receptor (AT1) antagonist and neprilysin (NEP) inhibitor with a pKi of 8 9 for AT1 and a pIC50 of 9 2 for NEP[1]
Catalog No. 50-002-30011 Supplier Medchemexpress LLC Supplier No. HY114412A10MM 1ML
TD-0212 TFA is an orally active dual pharmacology angiotensin II type 1 receptor (AT1) antagonist and neprilysin (NEP) inhibitor with a pKi of 8 9 for AT1 and a pIC50 of 9 2 for NEP[1]