Learn More
Medchemexpress LLC HY-114412A 5mg Medchemexpress, TD-0212 (TFA) CAS:1073549-11-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY114412A5MG
Encompass_Preferred
Medchemexpress, HY-114412A 5mg TD-0212 (TFA) CAS:1073549-11-7 TD-0212 TFA is an orally active dual pharmacology angiotensin II type 1 receptor (AT1) antagonist and neprilysin (NEP) inhibitor, with a pKi of 8.9 for AT1 and a pIC50 of 9.2 for NEP. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9351