Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC TD-106 100mg | 2250288-69-6 | 100 MG
SDP

Supplier:  Medchemexpress LLC HY114406100MG

Encompass_Preferred

TD-106 is a cereblon (CRBN) modulator used as an E3 ligase ligand in targeted protein degradation research. It has molecular formula C12H11N5O3 and a molecular weight of 273.25 g/mol. The compound is supplied at high reported purity for use in in vitro research, and BRD4-targeting PROTACs incorporating this ligand have been reported to induce BRD4 degradation.

  • Cereblon (CRBN) modulator used in PROTAC research.
  • Molecular formula C12H11N5O3.
  • Molecular weight 273.25 g/mol.
  • Reported purity 99.81%.
  • Available in multiple pack sizes, including 100 mg.
  • For research use only.

Catalog No. 50-002-26946


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.