Learn More
Medchemexpress LLC TD-106 25mg | 2250288-69-6 | 25 MG
SDP
Catalog No. 5000230045
Encompass_Preferred
TD-106 is a cereblon (CRBN) modulator used as an E3 ligase ligand in targeted protein degradation research, including BRD4-directed PROTAC studies.
  • Used as an E3 ligase ligand for PROTAC development and degradation assays.
  • Demonstrated to induce BRD4 degradation in cellular studies.
  • Provided at high purity appropriate for biochemical and cellular research.
  • Available in milligram-scale pack sizes for research workflows.
Catalog No. 50-002-30045 Supplier Medchemexpress LLC Supplier No. HY11440625MG
TD-106 is a cereblon (CRBN) modulator used as an E3 ligase ligand in targeted protein degradation research, including BRD4-directed PROTAC studies.
  • Used as an E3 ligase ligand for PROTAC development and degradation assays.
  • Demonstrated to induce BRD4 degradation in cellular studies.
  • Provided at high purity appropriate for biochemical and cellular research.
  • Available in milligram-scale pack sizes for research workflows.