Learn More
Medchemexpress LLC HY-114406 5mg Medchemexpress, TD-106 CAS:2250288-69-6 Purity:>98%
SDP
Catalog No. 501932487
Encompass_Preferred
Medchemexpress, HY-114406 5mg TD-106 CAS:2250288-69-6 TD-106 is a cereblon ( CRBN ) modulator, which can be used for targeted protein degradation. BRD4 PROTAC s with TD-106 induce BRD4 degradation [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2487 Supplier Medchemexpress LLC Supplier No. HY1144065MG
Medchemexpress, HY-114406 5mg TD-106 CAS:2250288-69-6 TD-106 is a cereblon ( CRBN ) modulator, which can be used for targeted protein degradation. BRD4 PROTAC s with TD-106 induce BRD4 degradation [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.