Learn More
Selleck Chemical LLC TD-139 5mg
SDP
Catalog No. 502173038
Encompass_Preferred
TD-139 (GB0-139) is a potent and selective inhibitor ofgalectin-3 with Kd of 0.036 ¿¿ over galectin-1 and galectin-7 with Kd of 2.2 ¿¿ and 32 ¿¿, respectively. TD-139 is used potentially for the treatment of idiopathic pulmonary fibrosis (IP *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3038 Supplier Selleck Chemical LLC Supplier No. S04715MG
TD-139 (GB0-139) is a potent and selective inhibitor ofgalectin-3 with Kd of 0.036 ¿¿ over galectin-1 and galectin-7 with Kd of 2.2 ¿¿ and 32 ¿¿, respectively. TD-139 is used potentially for the treatment of idiopathic pulmonary fibrosis (IP *For Research & Development use only.  Product is not intended for drug, household, or human consumption.