Learn More
Abcam TD-139, 1MG
Supplier:  Abcam AB2869721MG
Encompass_Preferred
MW 648.6 Da, Purity >=98%. A potent, synthetic inhibitor of galectin-3. TD-139 has high affinity for galectin-3 with a Kd of 14 nM and 10 nM for galectin-1, but low affinity for galectins 2, 4N, 4C, 7, 8N, or 9N.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-220-5823