Learn More
Chemglass Life Sciences Chemglass PVC Laboratory Standard Wall Tubing, Size 14 - 3/8 in ID x 5/8 in OD x 1/8 in wall, 50 ft roll
SDP
Catalog No. NC2583404
Encompass_Preferred
Chemglass PVC laboratory standard wall tubing is a flexible, durable PVC tubing designed for laboratory transfer of liquids and gases. It features a smooth, crystal-clear inner bore for easy visual monitoring, reduced buildup, and simplified cleaning, and it is formulated to withstand autoclaving and repeated gamma sterilization.
  • Flexible and durable for repeated use.
  • Smooth, clear inner bore reduces buildup and simplifies cleaning.
  • Resists autoclaving and repeated gamma sterilization.
  • 3/8 in ID x 5/8 in OD x 1/8 in wall; 50 ft per roll.
  • Suitable for transfer of liquids and gases in laboratory settings.
Catalog No. NC2583404 Supplier Chemglass Life Sciences Supplier No. CG730014
Chemglass PVC laboratory standard wall tubing is a flexible, durable PVC tubing designed for laboratory transfer of liquids and gases. It features a smooth, crystal-clear inner bore for easy visual monitoring, reduced buildup, and simplified cleaning, and it is formulated to withstand autoclaving and repeated gamma sterilization.
  • Flexible and durable for repeated use.
  • Smooth, clear inner bore reduces buildup and simplifies cleaning.
  • Resists autoclaving and repeated gamma sterilization.
  • 3/8 in ID x 5/8 in OD x 1/8 in wall; 50 ft per roll.
  • Suitable for transfer of liquids and gases in laboratory settings.