Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC TD52 dihydrochloride | 00-00-0 | 99.6% | 433.33 g/mol | C24H18Cl2N4 | 5 MG
SDP

Catalog No. 5000223329
Encompass_Preferred

TD52 dihydrochloride is a research-grade small-molecule inhibitor of cancerous inhibitor of protein phosphatase 2A (CIP2A). An Erlotinib derivative, it is orally active and induces apoptosis in triple-negative breast cancer cells by modulating the CIP2A/PP2A/p-Akt signaling pathway. Supplied as the dihydrochloride salt, the compound includes an alkyne group that enables click-chemistry applications.

  • High purity suitable for research applications (99.6%).
  • Available in small research pack sizes, including 5 mg.
  • Supplied as solid powder and as a 10 mM solution in DMSO.
  • Targeted for oncology and cell-signaling studies involving CIP2A/PP2A.
  • Compatible with copper-catalyzed azide-alkyne cycloaddition reactions.
  • Molecular formula C24H18Cl2N4; molecular weight 433.33 g/mol.

Catalog No. 50-002-23329 Supplier Medchemexpress LLC Supplier No. HY135699A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TD52 dihydrochloride is a research-grade small-molecule inhibitor of cancerous inhibitor of protein phosphatase 2A (CIP2A). An Erlotinib derivative, it is orally active and induces apoptosis in triple-negative breast cancer cells by modulating the CIP2A/PP2A/p-Akt signaling pathway. Supplied as the dihydrochloride salt, the compound includes an alkyne group that enables click-chemistry applications.

  • High purity suitable for research applications (99.6%).
  • Available in small research pack sizes, including 5 mg.
  • Supplied as solid powder and as a 10 mM solution in DMSO.
  • Targeted for oncology and cell-signaling studies involving CIP2A/PP2A.
  • Compatible with copper-catalyzed azide-alkyne cycloaddition reactions.
  • Molecular formula C24H18Cl2N4; molecular weight 433.33 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.