Learn More
Medchemexpress LLC Td52 dihydrochloride | 00-00-0 | 99.6% | C24H18Cl2N4 | 10MG
SDP
Catalog No. 5000259198
Encompass_Preferred
TD52 dihydrochloride is an Erlotinib-derived small-molecule inhibitor of CIP2A that induces apoptosis through the CIP2A/PP2A/p-Akt signaling pathway and demonstrates oral activity in cellular models. The compound contains an alkyne group enabling copper-catalyzed azide-alkyne cycloaddition for click-chemistry applications and is supplied as a high-purity solid with defined storage recommendations.
  • CIP2A inhibitor with apoptotic activity in cancer cells.
  • Erlotinib-derived scaffold with demonstrated oral activity in models.
  • Contains an alkyne for click chemistry applications (CuAAC).
  • High purity (≈99.6%) solid, light yellow to yellow appearance.
  • Stable with specified storage conditions for powder and solutions.
Catalog No. 50-002-59198 Supplier Medchemexpress LLC Supplier No. HY135699A10MG
TD52 dihydrochloride is an Erlotinib-derived small-molecule inhibitor of CIP2A that induces apoptosis through the CIP2A/PP2A/p-Akt signaling pathway and demonstrates oral activity in cellular models. The compound contains an alkyne group enabling copper-catalyzed azide-alkyne cycloaddition for click-chemistry applications and is supplied as a high-purity solid with defined storage recommendations.
  • CIP2A inhibitor with apoptotic activity in cancer cells.
  • Erlotinib-derived scaffold with demonstrated oral activity in models.
  • Contains an alkyne for click chemistry applications (CuAAC).
  • High purity (≈99.6%) solid, light yellow to yellow appearance.
  • Stable with specified storage conditions for powder and solutions.