Learn More
Cayman Chemical TDFAtrIfluoroacetate salt 10mg
SDP
Catalog No. 502964287
Encompass_Preferred
An inhibitor of JNK (IC50 280 nM) competitive for JNK1 binding with pepJIP1 (IC50 500 nM) selective for JNK over p38a mTOR and PI3Ka inhibits c-Jun phosphorylation induced by TNF-a (EC50 12 4 UM) restores insulin sensitivity in a mouse model of type 2 diabetes when administered at 25 mg/kg 30 minutes prior to insulin reduces human prostate strip contractions induced by phenylephrine or norepinephrine when applied at 30 UM and reduces phosphorylation of c-Jun a JNK substrate
Catalog No. 50-296-4287 Supplier Cayman Chemical Supplier No. 2248410 MG
An inhibitor of JNK (IC50 280 nM) competitive for JNK1 binding with pepJIP1 (IC50 500 nM) selective for JNK over p38a mTOR and PI3Ka inhibits c-Jun phosphorylation induced by TNF-a (EC50 12 4 UM) restores insulin sensitivity in a mouse model of type 2 diabetes when administered at 25 mg/kg 30 minutes prior to insulin reduces human prostate strip contractions induced by phenylephrine or norepinephrine when applied at 30 UM and reduces phosphorylation of c-Jun a JNK substrate