Learn More
Cayman Chemical TDG Thymine DNA glycosylase
SDP
Catalog No. 502947764
Encompass_Preferred
A mixture of porcine ganglioside GT1b with primarily C180 fatty acyl chain lengths as well as a lower amount of C200 fatty acyl chain lengths among various others characterized by having two sialic residues linked to the inner galactose unit binds to the neurotoxins BTxA BTxA heavy chain and tetanus toxin (IC50s 11 0.74 and 7.2 M respectively) ganglioside GT1b-containing liposomes can bind to the major coat protein VP1 from MCPyV a putative MCPyV receptor decreases spontaneous production of IL-6 IL-10 IgG IgM and IgA in human PBMCs by 31.4 30.5 60 59.5 and 58 respectively at 10 M
Catalog No. 50-294-7764 Supplier Cayman Chemical Supplier No. 32981100 UL
A mixture of porcine ganglioside GT1b with primarily C180 fatty acyl chain lengths as well as a lower amount of C200 fatty acyl chain lengths among various others characterized by having two sialic residues linked to the inner galactose unit binds to the neurotoxins BTxA BTxA heavy chain and tetanus toxin (IC50s 11 0.74 and 7.2 M respectively) ganglioside GT1b-containing liposomes can bind to the major coat protein VP1 from MCPyV a putative MCPyV receptor decreases spontaneous production of IL-6 IL-10 IgG IgM and IgA in human PBMCs by 31.4 30.5 60 59.5 and 58 respectively at 10 M