Learn More
Aviva Systems Biology Corp TDGF1 Antibody - middle region (ARP45440_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45440_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TDGF1. It was validated on Western Blot. TDGF1,teratocarcinoma-derived growth factor 1,CR; CRGF; CRIPTO; TDGF1,teratocarcinoma-derived growth factor 1,teratocarcinoma-derived growth factor 1,cripto-1 growth factor,epidermal growth factor-like cripto protein CR1The immunogen is a synthetic peptide directed towards the middle region of human TDGF1. Alternate names: CR, CRGF, CRIPTO, Cripto-1ApplicationWB. Species reactivity: Human
Catalog No. 50-285-200