Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC tDHU, acid | 2377643-37-1 | 98.8% | 248.23 g/mol | C12H12N2O4 | 50 MG
SDP

Catalog No. 5000281405
Encompass_Preferred

tDHU, acid is a dihydropyrimidine cereblon ligand developed as an E3 ubiquitin ligase ligand-linker conjugate for PROTAC development. It includes a benzoic acid functional handle for conjugation and is provided as a solid for medicinal chemistry and chemical biology research.

  • Designed as an E3 ligase ligand-linker conjugate for PROTAC development.
  • Contains a benzoic acid functional handle for conjugation.
  • High purity (98.8%) suitable for research applications.
  • Molecular weight 248.23 g/mol; formula C12H12N2O4.
  • White to off-white solid appearance; powder storage stable at -20°C.
  • Available in small-quantity packages for medicinal chemistry use.

Catalog No. 50-002-81405 Supplier Medchemexpress LLC Supplier No. HYW45490650MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

tDHU, acid is a dihydropyrimidine cereblon ligand developed as an E3 ubiquitin ligase ligand-linker conjugate for PROTAC development. It includes a benzoic acid functional handle for conjugation and is provided as a solid for medicinal chemistry and chemical biology research.

  • Designed as an E3 ligase ligand-linker conjugate for PROTAC development.
  • Contains a benzoic acid functional handle for conjugation.
  • High purity (98.8%) suitable for research applications.
  • Molecular weight 248.23 g/mol; formula C12H12N2O4.
  • White to off-white solid appearance; powder storage stable at -20°C.
  • Available in small-quantity packages for medicinal chemistry use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.