Learn More
TARGETMOL CHEMICALS INC TDI-011536 5MG
SDP
Catalog No. 502579666
Encompass_Preferred
Also available in 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. TDI-011536 is a potent Lats kinase inhibitor. TDI-011536 interrupts Hippo-Yap signaling and initiates the proliferation of lesioned heartmuscle cells. TDI-011536 can be used in studies of organ conservation and regeneration. purity: 98%
Catalog No. 50-257-9666 Supplier TARGETMOL CHEMICALS INC Supplier No. T601445MG
Also available in 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. TDI-011536 is a potent Lats kinase inhibitor. TDI-011536 interrupts Hippo-Yap signaling and initiates the proliferation of lesioned heartmuscle cells. TDI-011536 can be used in studies of organ conservation and regeneration. purity: 98%