Learn More
Selleck Chemical LLC TDI-011536-E1314-5MG
SDP
Supplier:  Selleck Chemical LLC E13145MG
Encompass_Preferred
TDI-011536 is a potent Lats kinase inhibitor that interrupts Hippo-Yap signaling and promotes proliferation of lesioned cardiac muscle cells
Catalog No. 50-239-0041