Learn More
Medchemexpress LLC TDI-10229 | 2810887-45-5 | 99.8% | C16H16ClN5 | 10MG
SDP
Catalog No. 5000261048
Encompass_Preferred
TDI-10229 is a small-molecule research inhibitor of soluble adenylyl cyclase (sAC, ADCY10) that displays nanomolar potency in biochemical and cellular assays (reported IC50 ≈195 nM). Supplied as a high-purity solid, it is intended for in vitro biochemical assays, cell-based studies, and medicinal chemistry research focused on cAMP signaling pathways.
  • High purity (~99.8%) suitable for sensitive assays.
  • Documented sAC inhibitory activity (IC50 ≈195 nM).
  • Supplied as a stable solid with recommended cold storage for long-term stability.
  • Appropriate for biochemical and cellular assay applications.
Catalog No. 50-002-61048 Supplier Medchemexpress LLC Supplier No. HY13229810MG
TDI-10229 is a small-molecule research inhibitor of soluble adenylyl cyclase (sAC, ADCY10) that displays nanomolar potency in biochemical and cellular assays (reported IC50 ≈195 nM). Supplied as a high-purity solid, it is intended for in vitro biochemical assays, cell-based studies, and medicinal chemistry research focused on cAMP signaling pathways.
  • High purity (~99.8%) suitable for sensitive assays.
  • Documented sAC inhibitory activity (IC50 ≈195 nM).
  • Supplied as a stable solid with recommended cold storage for long-term stability.
  • Appropriate for biochemical and cellular assay applications.