Learn More
TARGETMOL CHEMICALS INC TDI-10229 5MG
SDP
Catalog No. 502579371
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. TDI-10229 is a potent and orally available inhibitor of soluble adenylyl cyclase (sAC ADCY10). TDI-10229 displays nanomolar inhibition of sAC in both biochemical and cellular assays (IC50= 195 nM). TDI-10229 exhibits mouse pharmacokinetic properties sufficient to warrant its use as an in vivo tool compound. purity: 97%
Catalog No. 50-257-9371 Supplier TARGETMOL CHEMICALS INC Supplier No. T643555MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. TDI-10229 is a potent and orally available inhibitor of soluble adenylyl cyclase (sAC ADCY10). TDI-10229 displays nanomolar inhibition of sAC in both biochemical and cellular assays (IC50= 195 nM). TDI-10229 exhibits mouse pharmacokinetic properties sufficient to warrant its use as an in vivo tool compound. purity: 97%