Learn More
Medchemexpress LLC HY-132298 5mg , TDI-10229 CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1322985MG
Encompass_Preferred
HY-132298 5mg TDI-10229 CAS: TDI-10229 is a potent and orally bioavailable inhibitor of soluble adenylyl cyclase (sAC, ADCY10). TDI-10229 displays nanomolar inhibition of sAC in both biochemical and cellular assays (IC50 of 195 nM) and exhibits mouse pharmacokinetic properties sufficient to warrant its use as an in vivo tool compound. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1371